Learn More
Biomatik Corporation Phospho-MSK1 (Ser212) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51633
Encompass_Preferred
Phospho-MSK1 (Ser212) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MSK1 (Ser212) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75582. Gene ID: 9252. Target Synonyms: Nuclear mitogen- and stress-activated protein kinase 1; 90 kDa ribosomal protein S6 kinase 5; RSK-like protein kinase; RSKL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1747