Learn More
Biomatik Corporation Phospho-Src (Tyr216) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51657
Encompass_Preferred
Phospho-Src (Tyr216) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Src (Tyr216) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12931. Gene ID: 6714. Target Synonyms: c-Src; C-SRC; kinase Src; Neuronal SRC; p60-SRC; p60-Src; Src; SRC; SRC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1766