Learn More
Biomatik Corporation Phospho-Claudin 7 (Tyr210) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51671
Encompass_Preferred
Phospho-Claudin 7 (Tyr210) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Claudin 7 (Tyr210) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95471. Gene ID: 1366. Target Synonyms: CEPTRL2; claudin 7; claudin 9; claudin-1; claudin-7; CLD7; CLDN-7; CLDN7; clostridium perfringens enterotoxin receptor-like 2; CPETRL2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1779