Learn More
Biomatik Corporation Phospho-PKC ε (Ser729) Colorimetric Cell-Based ELISA Kit
Catalog No. 502289035
Encompass_Preferred
Phospho-PKC ε (Ser729) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKC ε (Ser729) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02156. Gene ID: 5581. Target Synonyms: KPCE; PKC-epsilon; PKCEA; PRKCE; Protein kinase C; epsilon type; kinase PKC-epsilon; nPKC-epsilon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9035 Supplier Biomatik Corporation Supplier No. EKA51241
Phospho-PKC ε (Ser729) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKC ε (Ser729) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02156. Gene ID: 5581. Target Synonyms: KPCE; PKC-epsilon; PKCEA; PRKCE; Protein kinase C; epsilon type; kinase PKC-epsilon; nPKC-epsilon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.