Learn More
Biomatik Corporation Phospho-HP1γ (Ser93) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51656
Encompass_Preferred
Phospho-HP1γ (Ser93) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HP1γ (Ser93) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13185. Gene ID: 11335; 653972. Target Synonyms: chromobox 3; chromobox 3 (HP1 gamma) HECH; heterochromatin protein 1 gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HP1 gamma; HP1 gamma homolog; HP1-GAMMA; HP1Hs-gamma; modifier 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1765