Learn More
Biomatik Corporation Phospho-S6 Ribosomal Protein (Ser235+Ser236) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51686
Encompass_Preferred
Phospho-S6 Ribosomal Protein (Ser235+Ser236) Colorimetric Cell-Based ELISA Kit for detecting Phospho-S6 Ribosomal Protein (Ser235+Ser236) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P62753. Gene ID: 6194. Target Synonyms: 40S ribosomal protein S6; Phosphoprotein NP33; ribosomal protein S6; RPS6; RS6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1794