Learn More
Biomatik Corporation Phospho-TOP2A (Ser1525) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51690
Encompass_Preferred
Phospho-TOP2A (Ser1525) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TOP2A (Ser1525) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11388. Gene ID: 7153. Target Synonyms: alpha isozyme; DNA topoisomerase II; DNA topoisomerase II-alpha; P11388-1; TOP-2; TOP2; TOP2A; TP2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1798