Learn More
Biomatik Corporation Phospho-FADD (Ser191) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51697
Encompass_Preferred
Phospho-FADD (Ser191) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FADD (Ser191) in researchers plate their cell line of choice. Species Reactivity: Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q61160. Gene ID: 14082. Target Synonyms: FAS-associating death domain-containing protein; Mediator of receptor induced toxicity; MORT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1801