Learn More
Biomatik Corporation Phospho-Integrin β1 (Tyr795) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291804
Encompass_Preferred
Phospho-Integrin β1 (Tyr795) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Integrin β1 (Tyr795) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05556. Gene ID: 3688. Target Synonyms: CD29; Fibronectin receptor beta subunit; FNRB; integrin beta-1; Integrin beta-1 precursor; Integrin VLA-4 beta subunit; ITB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1804 Supplier Biomatik Corporation Supplier No. EKA51700
Phospho-Integrin β1 (Tyr795) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Integrin β1 (Tyr795) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05556. Gene ID: 3688. Target Synonyms: CD29; Fibronectin receptor beta subunit; FNRB; integrin beta-1; Integrin beta-1 precursor; Integrin VLA-4 beta subunit; ITB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.