Learn More
Biomatik Corporation Phospho-MAP3K7 (Thr184) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51353
Encompass_Preferred
Phospho-MAP3K7 (Thr184) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAP3K7 (Thr184) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43318. Gene ID: 6885. Target Synonyms: M3K7; MAP3K7; Mitogen-activated protein kinase kinase kinase 7; TGF-beta- activated kinase 1; Transforming growth factor-beta-activated kinase 1; kinase TAK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1516