Learn More
Biomatik Corporation Phospho-SSB (Ser366) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51370
Encompass_Preferred
Phospho-SSB (Ser366) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SSB (Ser366) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05455. Gene ID: 6741. Target Synonyms: Lupus La protein; Sjoegren syndrome type B antigen; SS-B; La ribonucleoprotein; La autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1530