Learn More
Biomatik Corporation Phospho-CDK5 (Tyr15) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51358
Encompass_Preferred
Phospho-CDK5 (Tyr15) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDK5 (Tyr15) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q00535. Gene ID: 1020. Target Synonyms: CDK5; Cell division protein kinase 5; CRK6; kinase Cdk5; PSSALRE; Tau protein kinase II catalytic subunit; TAU protein kinase II catalytic subunit; TPKII catalytic subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9045