Learn More
Biomatik Corporation Phospho-ASK1 (Ser83) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51393
Encompass_Preferred
Phospho-ASK1 (Ser83) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ASK1 (Ser83) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99683. Gene ID: 4217. Target Synonyms: Apoptosis signal-regulating kinase 1; ASK-1; kinase ASK1; M3K5; MAP3K5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; MEKK5; Mitogen-activated protein kinase kinase kinase 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1550