Learn More
Biomatik Corporation Phospho-c-Jun (Ser63) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51418
Encompass_Preferred
Phospho-c-Jun (Ser63) Colorimetric Cell-Based ELISA Kit for detecting Phospho-c-Jun (Ser63) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05412. Gene ID: 3725. Target Synonyms: Activator protein 1; AH119; AP-1; AP1; c-Jun; Jun A; p39; Proto-oncogene c-jun; RJG-9; V-jun avian sarcoma virus 17 oncogene homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1569