Learn More
Biomatik Corporation Phospho-Connexin 43 (Ser367) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51421
Encompass_Preferred
Phospho-Connexin 43 (Ser367) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Connexin 43 (Ser367) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17302. Gene ID: 2697. Target Synonyms: Connexin 43; CX43; CXA1; CXN-43; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; GJA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1571