Learn More
Biomatik Corporation Phospho-p62 Dok (Tyr398) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291577
Encompass_Preferred
Phospho-p62 Dok (Tyr398) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p62 Dok (Tyr398) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99704. Gene ID: 1796. Target Synonyms: Docking protein 1; Downstream of tyrosine kinase 1; p62(dok) pp62; DOK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1577 Supplier Biomatik Corporation Supplier No. EKA51429
Phospho-p62 Dok (Tyr398) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p62 Dok (Tyr398) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99704. Gene ID: 1796. Target Synonyms: Docking protein 1; Downstream of tyrosine kinase 1; p62(dok) pp62; DOK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.