Learn More
Biomatik Corporation Phospho-Estrogen Receptor-α (Ser167) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291591
Encompass_Preferred
Phospho-Estrogen Receptor-α (Ser167) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Estrogen Receptor-α (Ser167) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03372. Gene ID: 2099. Target Synonyms: ER; ESR; ESR1; ESTR; ESTRA; Estradiol receptor; Estrogen receptor; NR3A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1591 Supplier Biomatik Corporation Supplier No. EKA51447
Phospho-Estrogen Receptor-α (Ser167) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Estrogen Receptor-α (Ser167) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03372. Gene ID: 2099. Target Synonyms: ER; ESR; ESR1; ESTR; ESTRA; Estradiol receptor; Estrogen receptor; NR3A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.