Learn More
Biomatik Corporation Phospho-G3BP-1 (Ser232) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51457
Encompass_Preferred
Phospho-G3BP-1 (Ser232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-G3BP-1 (Ser232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13283. Gene ID: 10146. Target Synonyms: Ras GTPase-activating protein-binding protein 1; G3BP-1; ATP-dependent DNA helicase VIII; HDH-VIII; GAP SH3 domain-binding protein 1; G3BP1; G3BP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1598