Learn More
Biomatik Corporation Phospho-LIMK1/2 (Thr508) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51497
Encompass_Preferred
Phospho-LIMK1/2 (Thr508) Colorimetric Cell-Based ELISA Kit for detecting Phospho-LIMK1/2 (Thr508) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P53667. Gene ID: 3984. Target Synonyms: kinase LIMK1; KIZ-1; LIM domain kinase 1; LIMK; LIMK-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1634