Learn More
Biomatik Corporation Phospho-MARCKS (Ser158) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51499
Encompass_Preferred
Phospho-MARCKS (Ser158) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MARCKS (Ser158) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29966. Gene ID: 4082. Target Synonyms: 80K-L protein; MACS; MARCS; myristoylated alanine-rich C-kinase substrate; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein light chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1636