Learn More
Biomatik Corporation Phospho-p38 MAPK (Thr180) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51530
Encompass_Preferred
Phospho-p38 MAPK (Thr180) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p38 MAPK (Thr180) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16539. Gene ID: 1432. Target Synonyms: protein; kinase p38-alpha; MAP kinase MXI2; MAP kinase p38; MAPK14; MAX-interacting protein 2; Mitogen-activated protein kinase 14; Mitogen-activated protein kinase p38; MK14; MXI2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1664