Learn More
Biomatik Corporation Phospho-PTEN (Ser380/Thr382/Thr383) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51551
Encompass_Preferred
Phospho-PTEN (Ser380/Thr382/Thr383) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PTEN (Ser380/Thr382/Thr383) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P60484. Gene ID: 5728. Target Synonyms: MMAC1; Mutated in multiple advanced cancers 1; phosphatase and tensin homolog gene; Protein-tyrosine phosphatase PTEN; TEP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1682