Learn More
Biomatik Corporation Phospho-STAT1 (Ser727) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51565
Encompass_Preferred
Phospho-STAT1 (Ser727) Colorimetric Cell-Based ELISA Kit for detecting Phospho-STAT1 (Ser727) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42224. Gene ID: 6772. Target Synonyms: Signal transducer and activator of transcription 1-alpha/beta; Transcription factor ISGF-3 components p91/p84. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1694