Learn More
Biomatik Corporation Phospho-AKT1/3 (Tyr437/434) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291735
Encompass_Preferred
Phospho-AKT1/3 (Tyr437/434) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AKT1/3 (Tyr437/434) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P31749; Q9Y243. Gene ID: 207; 10000. Target Synonyms: RAC-PK-alpha; Protein kinase B; PKB; C-AKT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1735 Supplier Biomatik Corporation Supplier No. EKA51618
Phospho-AKT1/3 (Tyr437/434) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AKT1/3 (Tyr437/434) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P31749; Q9Y243. Gene ID: 207; 10000. Target Synonyms: RAC-PK-alpha; Protein kinase B; PKB; C-AKT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.