Learn More
Biomatik Corporation Phospho-BL-CAM (Tyr807) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51655
Encompass_Preferred
Phospho-BL-CAM (Tyr807) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BL-CAM (Tyr807) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P20273. Gene ID: 933. Target Synonyms: B-cell receptor CD22; B-lymphocyte cell adhesion molecule; BL-CAM; Leu-14; Siglec-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1764