Learn More
Biomatik Corporation Phospho-WWOX (Tyr33) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51662
Encompass_Preferred
Phospho-WWOX (Tyr33) Colorimetric Cell-Based ELISA Kit for detecting Phospho-WWOX (Tyr33) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96KM3. Gene ID: 51741. Target Synonyms: Aberrant WW domain-containing oxidoreductase; FOR; WOX1; WW domain-containing oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1770