Learn More
Biomatik Corporation Phospho-BRCA1 (Ser1457) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51668
Encompass_Preferred
Phospho-BRCA1 (Ser1457) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BRCA1 (Ser1457) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P38398. Gene ID: 672. Target Synonyms: BRCA1; Breast cancer type 1 susceptibility protein; familial breast/ovarian cancer gene 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1776