Learn More
Biomatik Corporation Phospho-Claudin 6 (Tyr219) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51670
Encompass_Preferred
Phospho-Claudin 6 (Tyr219) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Claudin 6 (Tyr219) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P56747. Gene ID: 9074. Target Synonyms: claudin 6; claudin-6; CLD6; Skullin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1778