Learn More
Biomatik Corporation Phospho-EPB41 (Tyr660/418) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51674
Encompass_Preferred
Phospho-EPB41 (Tyr660/418) Colorimetric Cell-Based ELISA Kit for detecting Phospho-EPB41 (Tyr660/418) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11171. Gene ID: 2035. Target Synonyms: 4.1R; 41; band 4.1; E41P; EPB4.1; erythrocyte membrane protein band 4.1; P4.1; protein 4.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1782