Learn More
Biomatik Corporation Phospho-SREBP-1 (Ser439) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51687
Encompass_Preferred
Phospho-SREBP-1 (Ser439) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SREBP-1 (Ser439) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P36956. Gene ID: 6720. Target Synonyms: SRBP1; SREBF1; SREBP1; sterol regulatory element-binding protein 1; sterol regulatory element-binding transcription factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1795