Learn More
Biomatik Corporation Phospho-Abl (Tyr393/412) Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA51703
Encompass_Preferred
Phospho-Abl (Tyr393/412) Colorimetric Cell-Based ELISA for detecting Phospho-Abl (Tyr393/412) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P00519. Gene ID: 25; 27. Target Synonyms: p150; c-ABL; Abelson murine leukemia viral oncogene homolog 1/ Abelson murine leukemia viral oncogene homolog 2; Abelson-related gene protein; Tyrosine kinase ARG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1807