Learn More
Biomatik Corporation Phospho-Histone H2A.X (Ser139) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51472
Encompass_Preferred
Phospho-Histone H2A.X (Ser139) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Histone H2A.X (Ser139) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16104. Gene ID: 3014. Target Synonyms: H2A.X; H2AFX; H2a/x; HIST5-2AX; Histone H2A.X. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9067