Learn More
Biomatik Corporation Phospho-CDK2 (Thr160) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51002
Encompass_Preferred
Phospho-CDK2 (Thr160) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDK2 (Thr160) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24941. Gene ID: 1017. Target Synonyms: Cell division protein kinase 2; kinase Cdk2; p33 protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9000