Learn More
Biomatik Corporation Phospho-PKA α/β CAT (Thr197) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51143
Encompass_Preferred
Phospho-PKA α/β CAT (Thr197) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKA α/β CAT (Thr197) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P17612; P22694; P22612. Gene ID: 5566; 5567. Target Synonyms: cAMP-dependent protein kinase catalytic subunit alpha; PKA C-alpha; PRKACA ; PKACA ; cAMP-dependent protein kinase catalytic subunit beta; PKA C-beta; PRKACB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9021