Learn More
Raybiotech Inc Human Phospho-SMAD5 (SER463/465) and Total SMAD5 ELISA Kit, 2-Pack
Supplier:  Raybiotech Inc PELSMAD5S463T2
Encompass_Preferred
RayBio Human Phospho-SMAD5 (SER463/465) and Total SMAD5 ELISA Kit. This assay semi-quantitatively measures phosphorylated SMAD5 (Ser463/465) and Total SMAD5 in lysate samples.
Catalog No. NC1756813