Learn More
Biomatik Corporation Phospho-Myosin regulatory light chain 2 (Ser18) Cell-Based ELISA Kit, 192 tests
Supplier:  Biomatik Corporation EKA51201
Encompass_Preferred
Phospho-Myosin regulatory light chain 2 (Ser18) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Myosin regulatory light chain 2 (Ser18) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24844. Gene ID: 3566. Target Synonyms: Myosin regulatory light polypeptide 9; 20 kDa myosin light chain; LC20; MLC-2C; Myosin RLC; Myosin regulatory light chain 2 smooth muscle isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; MYL9; MLC2; MRLC1; MYRL2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-6640