Learn More
Raybiotech Inc Human/Mouse/Rat Guanine nucleotide-binding protein G(s) subunit alpha EIA Kit.
Supplier:  Raybiotech Inc EIAGSA1
Encompass_Preferred
Optimized for serum, plasma and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8820