Learn More
Raybiotech Inc Human Aminoacylase-1/ACY1 ELISA Kit
Supplier:  Raybiotech Inc ELHACY11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.19 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Aminoacylase-1.
Catalog No. 50-194-8946