Learn More
Raybiotech Inc Human Aminopeptidase LRAP/ERAP2 ELISA Kit
Supplier:  Raybiotech Inc ELHAPEPLRAP1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.23 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Aminopeptidase LRAP.
Catalog No. 50-194-8986