Learn More
Raybiotech Inc Human C1S/Complement Component C1s ELISA Kit
Supplier:  Raybiotech Inc ELHC1S1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human C1S.
Catalog No. 50-194-9044