Learn More
Raybiotech Inc Human Carboxylesterase 1/CES1 ELISA Kit
Supplier:  Raybiotech Inc ELHCES11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Carboxylesterase 1.
Catalog No. 50-194-9112