Learn More
Raybiotech Inc Human cIAP-1/BIRC2/hIAP-2 ELISA Kit
Supplier:  Raybiotech Inc ELHCIAP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.65 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human cIAP-1.
Catalog No. 50-194-9122