Learn More
Raybiotech Inc Human Carnosine Dipeptidase-1/CNDP1 ELISA Kit
Supplier:  Raybiotech Inc ELHCNDP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Carnosine Dipeptidase-1.
Catalog No. 50-194-9133