Learn More
Raybiotech Inc Human Crossveinless-2/CV-2/BMPER ELISA Kit
Supplier:  Raybiotech Inc ELHCV21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.45 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Crossveinless-2.
Catalog No. 50-194-9164