Learn More
Raybiotech Inc Human Coagulation Factor XIV/Protein C ELISA Kit
Supplier:  Raybiotech Inc ELHCOAGFXIV1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.82 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Coagulation Factor XIV.
Catalog No. 50-194-9142