Learn More
Raybiotech Inc Human Cytokeratin 18/KRT18 ELISA Kit
Supplier:  Raybiotech Inc ELHCYT181
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 160 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cytokeratin 18.
Catalog No. 50-194-9177