Learn More
Raybiotech Inc Human FGF-4 ELISA Kit
Supplier:  Raybiotech Inc ELHFGF4CL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 50 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human FGF-4.
Catalog No. 50-194-9263