Learn More
Raybiotech Inc Human IL-18 R alpha/IL-1 R5 ELISA Kit
Supplier:  Raybiotech Inc ELHIL1R51
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-18 R alpha.
Catalog No. 50-194-9423