Learn More
Raybiotech Inc Human IL-1 Ra (IL-1 F3) ELISA Kit
Supplier:  Raybiotech Inc ELHIL1RACL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 100 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human IL-1ra.
Catalog No. 50-194-9426