Learn More
Raybiotech Inc Human Lp-PLA2/PLA2G7/PAF-AH ELISA Kit
Supplier:  Raybiotech Inc ELHLPPLA21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Lp-PLA2.
Catalog No. 50-194-9507